Skip to Content
Merck
All Photos(1)

Key Documents

W530228

Sigma-Aldrich

Canola oil

Certified organic (NOP)

Synonym(s):

Rapeseed oil

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12164502
grade:
Certified organic (NOP)
Halal
Kosher
food allergen:
no known allergens
Product W530228 is not currently sold in your country. Contact Technical Service

grade

Certified organic (NOP)
Halal
Kosher

origin

Switzerland origin

refractive index

n20/D 1.473

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

faint

Looking for similar products? Visit Product Comparison Guide

General description

Canola oil, a widely used vegetable oil, is obtained from canola seeds. It contains low levels of erucic acid and glucosinolates when compared to crude rapeseed oil. Canola oil is generally composed of high amounts of monounsaturated fatty acids, polyunsaturated fatty acids and low level of saturated fatty acids. It may also contain phytosterols and vitamin E.[1]

Legal Information

Certified organic by MOSA

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Canola: Chemistry, Production, Processing, and Utilization, 245-247 (2015)
Ask the doctor. I use a lot of canola oil in cooking, because it's heart-healthy. My husband has prostate cancer, and I just heard that canola oil might make it worse. Is that really true and should I stop using it for cooking?
Anthony L Komaroff
Harvard health letter, 37(11), 2-2 (2012-10-19)
Jiang Jiang et al.
Meat science, 93(3), 469-476 (2013-01-01)
Alkali (pH(12)) and acid (pH(1.5)) pH-treated soy protein isolates (SPI) were incorporated (0.25-0.75% protein) into sols of myofibrillar protein (MP, 3%, in 0.6 M NaCl at pH 6.25) with or without 0.1% microbial transglutaminase (TG) to investigate the potential as
Laziz Bouzidi et al.
Food & function, 4(1), 130-143 (2012-10-17)
Pure saturated triacylglycerols (TAGs) in canola oil were used as model systems to analyse oil loss in structured oil both from thermodynamic and kinetic perspectives. Two important parameters which effectively and predictively measure the relative propensity of a solid network
Nancy G Faith et al.
Journal of food protection, 75(3), 567-572 (2012-07-26)
In this study, we explored the use of vinegar, or vinegar and canola oil as a salad dressing, to reduce bacterial levels on spinach leaves. We found that incubation of spinach leaves with various types of vinegar substantially reduced the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service