Skip to Content
Merck
All Photos(1)

Key Documents

W530216

Sigma-Aldrich

Palm fruit oil

Certified organic (NOP)

Synonym(s):

Palm oil

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12164502
grade:
Certified organic (NOP)
Fragrance grade
Halal
biological source:
Elaeis guineensis
food allergen:
no known allergens

biological source

Elaeis guineensis

grade

Certified organic (NOP)
Fragrance grade
Halal

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009

origin

Colombia origin

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

fruity

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Palm fruit oil, a widely used vegetable oil, is obtained from the mesocarp of the palm fruit. It is mainly composed of palmitic acid and also unsaturated fatty acids such oleic and linoleic acid. Palm oil undergoes trans-esterification with methanol to form methyl ester of palm oil, also known as palm oil diesel (POD). POD is a promising candidate as a diesel substitute.

Legal Information

Certified organic by MOSA

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The use of palm oil as diesel fuel substitute.
Sapuan SM, et al.
Proceedings of the Institution of Mechanical Engineers, Part A: Journal of Power and Energy, 210(1), 47-53 (1996)
Biodiesel from palmoil?an analysis of its properties and potential.
Kalam MA & Masjuki HH.
Biomass and Bioenergy, 23(6), 471-479 (2002)
Chee-Meng Boon et al.
PloS one, 8(2), e55908-e55908 (2013-02-15)
Oleic acid has been shown to lower high blood pressure and provide cardiovascular protection. Curiosity arises as to whether super olein (SO), red palm olein (RPO) and palm olein (PO), which have high oleic acid content, are able to prevent
Khamphe Phoungthong et al.
Journal of environmental sciences (China), 25(4), 751-757 (2013-08-09)
Mixed crude palm oil (MCPO), the mixture of palm fiber oil and palm kernel oil, has become of great interest as a renewable energy source. It can be easily extracted from whole dried palm fruits. In the present work, the
Catherine A O Akinbami et al.
Ambio, 42(5), 644-657 (2013-01-24)
In rural Nigeria, food processing is mostly engaged in by women and children. Most of these processes are done using outdated technologies that make use of traditional woodstoves. This article presents the health and environmental implications of the rural female

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service