Skip to Content
Merck
All Photos(1)

Key Documents

W516104

Sigma-Aldrich

2-tert-Butyl-5-methylphenol

≥98%

Synonym(s):

6-tert-Butyl-m-cresol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CC6H3(CH3)OH
CAS Number:
Molecular Weight:
164.24
EC Number:
MDL number:
UNSPSC Code:
12164502
Flavis number:
4.078

Assay

≥98%

refractive index

n20/D 1.519 (lit.)

bp

117-118 °C/12 mmHg (lit.)

density

0.964 g/mL at 25 °C (lit.)

SMILES string

Cc1ccc(c(O)c1)C(C)(C)C

InChI

1S/C11H16O/c1-8-5-6-9(10(12)7-8)11(2,3)4/h5-7,12H,1-4H3

InChI key

XOUQAVYLRNOXDO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

222.8 °F - closed cup

Flash Point(C)

106 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service