Skip to Content
Merck
All Photos(1)

Key Documents

W502405

Sigma-Aldrich

1,2,6-Trihydroxyhexane

≥95%

Synonym(s):

1,2,6-Hexanetriol, 1,2,6-Trihydroxyhexane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO(CH2)4CH(OH)CH2OH
CAS Number:
Molecular Weight:
134.17
Beilstein:
1304479
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:

biological source

synthetic

Quality Level

grade

Kosher

vapor density

4.63 (vs air)

vapor pressure

<0.01 mmHg ( 20 °C)

Assay

≥95%

refractive index

n20/D 1.476 (lit.)

bp

178 °C/5 mmHg (lit.)

density

1.109 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Organoleptic

ethereal

SMILES string

OCCCCC(O)CO

InChI

1S/C6H14O3/c7-4-2-1-3-6(9)5-8/h6-9H,1-5H2

InChI key

ZWVMLYRJXORSEP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

383.0 °F - closed cup

Flash Point(C)

195 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service