Skip to Content
Merck
All Photos(1)

Documents

W501603

Sigma-Aldrich

3-Furancarboxaldehyde

≥97%

Synonym(s):

3-Furaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H4O2
CAS Number:
Molecular Weight:
96.08
Beilstein:
105852
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:

biological source

synthetic

Assay

≥97%

refractive index

n20/D 1.493 (lit.)

bp

144 °C/732 mmHg (lit.)

density

1.111 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

sulfurous

storage temp.

2-8°C

SMILES string

O=Cc1ccoc1

InChI

1S/C5H4O2/c6-3-5-1-2-7-4-5/h1-4H

InChI key

AZVSIHIBYRHSLB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

118.4 °F - closed cup

Flash Point(C)

48 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service