Skip to Content
Merck
All Photos(1)

Key Documents

W501018

Sigma-Aldrich

Isoamyl tiglate

≥97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=C(CH3)CO2CH2CH2CH(CH3)2
CAS Number:
Molecular Weight:
170.25
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
NACRES:
NA.21

biological source

synthetic

Quality Level

Assay

≥97%

refractive index

n20/D 1.441 (lit.)

bp

204-205 °C (lit.)

density

0.897 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fruity; herbaceous; wine-like

SMILES string

C\C=C(/C)C(=O)OCCC(C)C

InChI

1S/C10H18O2/c1-5-9(4)10(11)12-7-6-8(2)3/h5,8H,6-7H2,1-4H3/b9-5+

InChI key

ZARFDQHJMNVNLE-WEVVVXLNSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

176.0 °F - closed cup

Flash Point(C)

80 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sapir Penker et al.
Frontiers in systems neuroscience, 14, 604718-604718 (2020-12-18)
Coding of odorous stimuli has been mostly studied using single isolated stimuli. However, a single sniff of air in a natural environment is likely to introduce airborne chemicals emitted by multiple objects into the nose. The olfactory system is therefore

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service