Skip to Content
Merck
All Photos(1)

Key Documents

W479010

Sigma-Aldrich

Ethyl bicyclo[2.2.1]hept-2-ene-5-carboxylate

≥95%

Sign Into View Organizational & Contract Pricing

Select a Size

W479010-SAMPLE-K
$55.50
100 G
$139.00
1 KG
$1,340.00

$55.50


Check Cart for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
W479010-SAMPLE-K
$55.50
100 G
$139.00
1 KG
$1,340.00

About This Item

Empirical Formula (Hill Notation):
C10H14O2
CAS Number:
Molecular Weight:
166.22
FEMA Number:
4790
UNSPSC Code:
12164502
NACRES:
NA.21
grade:
Fragrance grade
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

$55.50


Check Cart for Availability

Request a Bulk OrderRequest more information

biological source

synthetic

Quality Level

grade

Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
FDA 21 CFR 117

Assay

≥95%

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

green; melon; fruity

SMILES string

O(CC)C(=O)C1C2CC(C1)C=C2

InChI

1S/C10H14O2/c1-2-12-10(11)9-6-7-3-4-8(9)5-7/h3-4,7-9H,2,5-6H2,1H3

InChI key

FCCGTJAGEHZPBF-UHFFFAOYSA-N

Related Categories

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service