Skip to Content
Merck
All Photos(1)

Key Documents

W467101

Sigma-Aldrich

(Z)-5-Octenyl acetate

≥95%

Synonym(s):

cis-5-Octenyl acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O2
CAS Number:
Molecular Weight:
170.25
FEMA Number:
4671
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
09.950
NACRES:
NA.21

Assay

≥95%

form

liquid

refractive index

n20/D 1.435

density

0.890 g/mL at 25 °C

application(s)

flavors and fragrances

Organoleptic

fatty; green; fruity; vegetable; tropical

storage temp.

2-8°C

SMILES string

CC(OCCCC/C=C\CC)=O

InChI

1S/C10H18O2/c1-3-4-5-6-7-8-9-12-10(2)11/h4-5H,3,6-9H2,1-2H3/b5-4-

InChI key

NBYQSRDPBAVRDT-PLNGDYQASA-N

General description

Odor Characteristics:@ 1.0% ETOH. Green, Fresh, Fatty, Slighty earthy vegetative green pepper like. Unripe apple, pear and melon with nuances of guava and kiwi fruit.
Taste Characteristics: @ 1 TO 2 PPM. Green fatty, earthy green pepper and lettuce vegetative. Unripe apple, Asian pear, banana with nuances of passion fruit, kiwi and Guava.

Application

This ingredient with its fatty green vegetative and unripe fruitiness would enhance green apple, pears, Asian pears and melon flavors. Its green vegetative notes would complement green pepper, lettuce and garden herbal notes. Its tropical nuances would freshen guava, passion fruit and kiwi flavors.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

178.0 °F

Flash Point(C)

81.1 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service