Skip to Content
Merck
All Photos(1)

Key Documents

W463901

Sigma-Aldrich

2-Methoxypyridine

≥98%

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$155.00
W463901-SAMPLE-K
$234.00
1 KG
$590.00

$155.00


Check Cart for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
100 G
$155.00
W463901-SAMPLE-K
$234.00
1 KG
$590.00

About This Item

Empirical Formula (Hill Notation):
C6H7NO
CAS Number:
Molecular Weight:
109.13
FEMA Number:
4639
Beilstein:
108189
MDL number:
UNSPSC Code:
12164502
NACRES:
NA.21
grade:
Halal
Kosher
food allergen:
no known allergens

$155.00


Check Cart for Availability

Request a Bulk OrderRequest more information

grade

Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

FDA 21 CFR 117

Assay

≥98%

refractive index

n20/D 1.503 (lit.)

bp

142 °C (lit.)

density

1.038 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

burnt; fatty; green

SMILES string

COc1ccccn1

InChI

1S/C6H7NO/c1-8-6-4-2-3-5-7-6/h2-5H,1H3

InChI key

IWTFOFMTUOBLHG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

89.6 °F - closed cup

Flash Point(C)

32 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service