Skip to Content
Merck
All Photos(1)

Key Documents

W416301

Sigma-Aldrich

2′-Methoxyacetophenone

99%

Synonym(s):

2-Acetylanisole

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4COCH3
CAS Number:
Molecular Weight:
150.17
FEMA Number:
4163
Beilstein:
1100602
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
7.254

biological source

synthetic

Assay

99%

refractive index

n20/D 1.5393 (lit.)

bp

131 °C/18 mmHg (lit.)

density

1.09 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Organoleptic

medicinal; earthy

SMILES string

COc1ccccc1C(C)=O

InChI

1S/C9H10O2/c1-7(10)8-5-3-4-6-9(8)11-2/h3-6H,1-2H3

InChI key

DWPLEOPKBWNPQV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Soluble in: Alcohol

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service