Skip to Content
Merck
All Photos(1)

Key Documents

W398311

Sigma-Aldrich

trans-2-Hexenyl hexanoate

≥93%, FG

Synonym(s):

2-Hexenyl hexanoate, (E)-2-Hexen-1-yl hexanoate

Sign Into View Organizational & Contract Pricing

Select a Size

W398311-SAMPLE
$75.00
100 G
$120.00
1 KG
$525.00

$75.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
W398311-SAMPLE
$75.00
100 G
$120.00
1 KG
$525.00

About This Item

Empirical Formula (Hill Notation):
C12H22O2
CAS Number:
Molecular Weight:
198.30
FEMA Number:
3983
MDL number:
UNSPSC Code:
12164502
Flavis number:
09.398
NACRES:
NA.21
grade:
FG
Halal
food allergen:
no known allergens

$75.00


Check Cart for Availability

Request a Bulk Order

grade

FG
Halal

Quality Level

reg. compliance

EU 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

≥93%

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

green; waxy; herbaceous

SMILES string

CCCCCC(=O)OC\C=C\CCC

InChI

1S/C12H22O2/c1-3-5-7-9-11-14-12(13)10-8-6-4-2/h7,9H,3-6,8,10-11H2,1-2H3/b9-7+

InChI key

UQPLEMTXCSYMEK-VQHVLOKHSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service