Skip to Content
Merck
All Photos(1)

Key Documents

W398000

Sigma-Aldrich

3-Heptyl acetate

natural (US), ≥96%, FG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H18O2
CAS Number:
Molecular Weight:
158.24
FEMA Number:
3980
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Flavis number:
09.924
grade:
FG
Halal
Kosher
natural (US)
food allergen:
no known allergens

grade

FG
Halal
Kosher
natural (US)

reg. compliance

EU Regulation 1334/2008 & 178/2002

vapor pressure

0.8625

Assay

≥96%

refractive index

n/D 1.410

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

green; fruity; pungent; floral; sweet

SMILES string

CCC(OC(C)=O)CCCC

InChI

1S/C9H18O2/c1-4-6-7-9(5-2)11-8(3)10/h9H,4-7H2,1-3H3

InChI key

QICIMADZMWGDTQ-UHFFFAOYSA-N

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

138.0 °F

Flash Point(C)

58.88 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service