Skip to Content
Merck
All Photos(1)

Key Documents

W396321

Sigma-Aldrich

2,3,6-Trimethylphenol

≥98%, FG

Sign Into View Organizational & Contract Pricing

Select a Size

W396321-SAMPLE
$67.20
100 G
$134.00
500 G
$471.00

$67.20


Check Cart for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
W396321-SAMPLE
$67.20
100 G
$134.00
500 G
$471.00

About This Item

Linear Formula:
(CH3)3C6H2OH
CAS Number:
Molecular Weight:
136.19
FEMA Number:
3963
Beilstein:
2042207
MDL number:
UNSPSC Code:
12352100
Flavis number:
04.085
NACRES:
NA.05
grade:
FG
food allergen:
no known allergens

$67.20


Check Cart for Availability

Request a Bulk OrderRequest more information

grade

FG

Quality Level

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

≥98%

mp

59-62 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

coffee; nutty; phenolic

SMILES string

Cc1ccc(C)c(O)c1C

InChI

1S/C9H12O/c1-6-4-5-7(2)9(10)8(6)3/h4-5,10H,1-3H3

InChI key

QQOMQLYQAXGHSU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service