Skip to Content
Merck
All Photos(1)

Documents

W394602

Sigma-Aldrich

2-Methylcyclohexanone

≥98%

Synonym(s):

Tetrahydro-o-cresol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H9(=O)
CAS Number:
Molecular Weight:
112.17
FEMA Number:
3946
Beilstein:
506751
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
7.179
NACRES:
NA.21

biological source

synthetic

Agency

meets purity specifications of JECFA

Assay

≥98%

refractive index

n20/D 1.448 (lit.)

bp

162-163 °C (lit.)

density

0.924 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

cooling; minty; peppermint

SMILES string

CC1CCCCC1=O

InChI

1S/C7H12O/c1-6-4-2-3-5-7(6)8/h6H,2-5H2,1H3

InChI key

LFSAPCRASZRSKS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Methylcyclohexanone is a cyclic ketone that is reported to occur in mint and horse chestnut.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

118.4 °F

Flash Point(C)

48 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Acaricidal potency of active constituent isolated from Mentha piperita and its structural analogs against pyroglyphid mites.
Lee HW & Lee HS.
Journal of the Korean Society for Applied Biological Chemistr, 58(4), 597-602 (2015)
Volatiles of common horsechestnut (Aesculus hippocastanum L.)(Hippocastanaceae) peels and seeds.
Buchbauer G, et al.
J. Essent. Oil Res., 6(5), 507-511 (1994)
Halis T Balaydın et al.
Bioorganic & medicinal chemistry letters, 22(3), 1352-1357 (2012-01-11)
The Naturally occurring novel cyclohexanonyl bromophenol 2(R)-2-(2,3,6-tribromo-4,5-dihydroxybenzyl)cyclohexanone (4) was synthesized as a racemic compound. Cyclohexylphenyl methane derivatives (10-17) with Br, OMe, CO, and OH were also obtained. Inhibition of four human carbonic anhydrase (hCA, EC 4.2.1.1) isozymes I, II, IV

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service