Skip to Content
Merck
All Photos(1)

Documents

W393011

Sigma-Aldrich

trans-2-Hexenyl isovalerate

≥95%, FG

Synonym(s):

2-Hexenyl 3-methylbutanoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H20O2
CAS Number:
Molecular Weight:
184.28
FEMA Number:
3930
MDL number:
UNSPSC Code:
12164502
Flavis number:
09.399
NACRES:
NA.21

grade

FG
Halal

Quality Level

reg. compliance

EU 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

≥95%

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

apple; banana; grape; green; waxy; pear

SMILES string

CCC\C=C\COC(=O)CC(C)C

InChI

1S/C11H20O2/c1-4-5-6-7-8-13-11(12)9-10(2)3/h6-7,10H,4-5,8-9H2,1-3H3/b7-6+

InChI key

SAVRWHQEMHIAEB-VOTSOKGWSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service