Skip to Content
Merck
All Photos(1)

Key Documents

W384208

Sigma-Aldrich

1-Hexanethiol

≥95%

Synonym(s):

1-Mercaptohexane, Hexyl mercaptan, MCH, Mercaptan C6

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5SH
CAS Number:
Molecular Weight:
118.24
FEMA Number:
3842
Beilstein:
1731295
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
12.132

biological source

synthetic

Assay

≥95%

refractive index

n20/D 1.450

bp

150-154 °C (lit.)

mp

−81-−80 °C (lit.)

density

0.832 g/mL at 25 °C

application(s)

flavors and fragrances

SMILES string

CCCCCCS

InChI

1S/C6H14S/c1-2-3-4-5-6-7/h7H,2-6H2,1H3

InChI key

PMBXCGGQNSVESQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Odor at 1%
Taste at 1-3 ppm

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

86.0 °F - closed cup

Flash Point(C)

30 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fallon Cook et al.
BMC pediatrics, 15, 174-174 (2015-11-12)
Anticipatory guidance around the management of sleep and crying problems in early infancy has been shown to improve both infant behaviour and parent symptoms of postnatal depression. Digital technology offers platforms for making such programs widely available in a cost-efficient
Simon Thebault et al.
PloS one, 10(6), e0129555-e0129555 (2015-06-20)
Type 1 narcolepsy is caused by deficiency of hypothalamic orexin/hypocretin. An autoimmune basis is suspected, but no specific antibodies, either causative or as biomarkers, have been identified. However, the AS03 adjuvanted split virion H1N1 (H1N1-AS03) vaccine, created to protect against
Chang Liu et al.
The Analyst, 139(21), 5460-5465 (2014-08-30)
Due to its low growth rate and its fastidious nature, Mycobacterium tuberculosis is difficult to identify. Its rapid and sensitive detection is, however, critical for the control of tuberculosis. Molecular biology, and more recently electrochemical technology, have been exploited for
Wei Wen et al.
Biosensors & bioelectronics, 71, 13-17 (2015-04-17)
In this work, a sensitive exonuclease-assisted amplification electrochemical aptasensor through insertion approach was developed for the detection of mucin 1 (MUC 1). In order to construct the aptasensor, 6-Mercapto-1-hexanol (MCH) was used to block partial sites of gold electrode (GE)
Siana Nkya Mtatiro et al.
American journal of hematology, 90(1), E1-E4 (2014-09-30)
Fetal hemoglobin (HbF) is a recognized modulator of sickle cell disease (SCD) severity. HbF levels are strongly influenced by genetic variants at three major genetic loci, Xmn1-HBG2, HMIP-2, and BCL11A, but the effect of these loci on the hematological phenotype

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service