Skip to Content
Merck
All Photos(1)

Key Documents

W379506

Sigma-Aldrich

1,3,5-Undecatriene, mixture of 1,3(E),5(Z) and 1,3(E),5(E) isomers

≥70%, stabilized

Synonym(s):

galbanum decatriene

Sign Into View Organizational & Contract Pricing

Select a Size

W379506-SAMPLE-K
$103.00
1 KG
$6,490.00

$103.00


Check Cart for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
W379506-SAMPLE-K
$103.00
1 KG
$6,490.00

About This Item

Linear Formula:
CH3(CH2)4(CH=CH)2CH=CH2
CAS Number:
Molecular Weight:
150.26
FEMA Number:
3795
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
1.061
NACRES:
NA.21
grade:
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

$103.00


Check Cart for Availability

Request a Bulk OrderRequest more information

biological source

synthetic

Quality Level

grade

Halal
Kosher

Assay

≥70%

contains

α-tocopherol, synthetic as stabilizer

refractive index

n20/D 1.513 (lit.)

bp

280-285 °C (lit.)

density

0.79 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fresh; green

SMILES string

CCCCC\C=C\C=C\C=C

InChI

1S/C11H18/c1-3-5-7-9-11-10-8-6-4-2/h3,5,7,9,11H,1,4,6,8,10H2,2H3/b7-5+,11-9+

InChI key

JQQDKNVOSLONRS-JEGFTUTRSA-N

Related Categories

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

172.4 °F - closed cup

Flash Point(C)

78 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service