Skip to Content
Merck
All Photos(2)

Key Documents

W376109

Sigma-Aldrich

5-Methyl-2-hepten-4-one solution

natural, 10 wt. % in ethanol, FG

Synonym(s):

Hazelnut Ketone

Sign Into View Organizational & Contract Pricing

Select a Size

W376109-SAMPLE-K
$97.10
25 G
$230.00
100 G
$623.00

$97.10


Check Cart for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
W376109-SAMPLE-K
$97.10
25 G
$230.00
100 G
$623.00

About This Item

Linear Formula:
C2H5CH(CH3)COCH=CHCH3
CAS Number:
Molecular Weight:
126.20
FEMA Number:
3761
Council of Europe no.:
2085
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
7.139
NACRES:
NA.21
grade:
FG
Kosher
natural
food allergen:
no known allergens

$97.10


Check Cart for Availability

Request a Bulk OrderRequest more information

grade

FG
Kosher
natural

Quality Level

reg. compliance

EU Regulation 1334/2008 & 178/2002

concentration

10 wt. % in ethanol

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

coffee; green; hazelnut; earthy

SMILES string

O=C(C(CC)C)\C=C\C

InChI

1S/C8H14O/c1-4-6-8(9)7(3)5-2/h4,6-7H,5H2,1-3H3/b6-4+

InChI key

ARJWAURHQDJJAC-GQCTYLIASA-N

General description

5-methyl-2-hepten-4-one (Filbertone) is one of the key aroma components in hazelnuts . It has a sweet, nutty, and slightly roasted aroma characteristic of hazelnuts [1]. In the food industry, filbertone is often used as a flavoring agent to simulate hazelnut flavor in various food products, including chocolate [2].

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

57.2 °F

Flash Point(C)

14 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comparison of natural and roasted Turkish tombul hazelnut (Corylus avellana L.) volatiles and flavor by DHA/GC/MS and descriptive sensory analysis
Alasalvar C, et al.
Journal of Agricultural and Food Chemistry, 51(7), 5067-5072 (2003)
Effect of in vitro digestion and fermentation on antioxidant capacity of weight loss foods and Maillard reaction products content
Perez-Burillo, S, et al.
Food Research International, 166, 112616-112616 (2023)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service