Skip to Content
Merck
All Photos(1)

Key Documents

W366404

Sigma-Aldrich

2,5-Dimethyl-4-methoxy-3(2H)-furanone

≥97%, FG

Synonym(s):

strawberry furanone methyl ether

Sign Into View Organizational & Contract Pricing

Select a Size

W366404-SAMPLE-K
$83.00
100 G
$297.00
1 KG
$1,930.00

$83.00


Check Cart for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
W366404-SAMPLE-K
$83.00
100 G
$297.00
1 KG
$1,930.00

About This Item

Empirical Formula (Hill Notation):
C7H10O3
CAS Number:
Molecular Weight:
142.15
FEMA Number:
3664
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
13.089
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

$83.00


Check Cart for Availability

Request a Bulk OrderRequest more information

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002

Assay

≥97%

refractive index

n20/D 1.478 (lit.)

density

1.097 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

caramel; coffee; maple; musty; sweet

storage temp.

2-8°C

SMILES string

COC1=C(C)OC(C)C1=O

InChI

1S/C7H10O3/c1-4-6(8)7(9-3)5(2)10-4/h4H,1-3H3

InChI key

SIMKGHMLPVDSJE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

2,5-Dimethyl-4-methoxy-3(2H)-furanone is one of the key volatile aroma compounds that contribute to the characteristic aroma of strawberries.[1][2][3]

Biochem/physiol Actions

Odor at 1.0% PG

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

167.0 °F - closed cup

Flash Point(C)

75 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Odour thresholds of some important aroma compounds in strawberries.
Larsen M & Poll L
World Journal of Gastroenterology, 195(2), 120-123 (1992)
2,5-Dimethyl-4-hydroxy-3(2H)-furanone and derivatives in strawberries during ripening.
Sanz C, et al.
ACS Symp. Ser. (1995)
Mosciano, G.
Perfumer & Flavorist, 24, 41-41 (1999)
Wilfried Schwab
Molecules (Basel, Switzerland), 18(6), 6936-6951 (2013-06-15)
4-Hydroxy-2,5-dimethyl-3(2H)-furanone (HDMF, furaneol®) and its methyl ether 2,5-dimethyl-4-methoxy-3(2H)-furanone (DMMF) are import aroma chemicals and are considered key flavor compounds in many fruit. Due to their attractive sensory properties they are highly appreciated by the food industry. In fruits 2,5-dimethyl-3(2H)-furanones are

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service