Skip to Content
Merck
All Photos(1)

Key Documents

W363421

Sigma-Aldrich

4,5-Dimethyl-3-hydroxy-2,5-dihydrofuran-2-one solution

3 wt. % in propylene glycol

Synonym(s):

Caramel furanone, Sotolon, Sugar lactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8O3
CAS Number:
Molecular Weight:
128.13
FEMA Number:
3634
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
10.030
NACRES:
NA.21
grade:
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

Halal
Kosher

concentration

3 wt. % in propylene glycol

refractive index

n20/D 1.434

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

caramel; maple; sweet

InChI

1S/C6H8O3/c1-3-4(2)9-6(8)5(3)7/h4,7H,1-2H3

InChI key

UNYNVICDCJHOPO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Natural occurrence: Fenugreek seed, roasted Virginia tobacco, sake, rice wine.

Biochem/physiol Actions

Odor at 1.0%
Taste at 5-25 ppm

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organoleptic Characteristics of Flavor Materials
Mosciano, G.
Perfumer & Flavorist, 30, 39-39 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service