Skip to Content
Merck
All Photos(1)

Key Documents

W336807

Sigma-Aldrich

4-Methyl-3-penten-2-one

90%

Synonym(s):

Mesityl oxide, 4-Methyl-3-penten-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C=CHCOCH3
CAS Number:
Molecular Weight:
98.14
FEMA Number:
3368
Beilstein:
1361550
EC Number:
Council of Europe no.:
11853
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
7.101
NACRES:
NA.21
Organoleptic:
earthy; vegetable
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

Assay

90%

impurities

≤10% 4-methyl-4-penten-2-one

refractive index

n20/D 1.442 (lit.)

bp

130 °C (lit.)

mp

-41.5 °C (lit.)

density

0.858 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

earthy; vegetable

SMILES string

C\C(C)=C\C(C)=O

InChI

1S/C6H10O/c1-5(2)4-6(3)7/h4H,1-3H3

InChI key

SHOJXDKTYKFBRD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

87.8 °F - closed cup

Flash Point(C)

31 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christian Starkenmann
Journal of agricultural and food chemistry, 51(24), 7146-7155 (2003-11-13)
Cysteine conjugates, resulting from the addition of cysteine to alpha,beta-unsaturated carbonyl compounds, are important precursors of odorant sulfur compounds in food flavors. The aim of this work was to better understand this chemistry in the light of the unexpected double
Z Jia et al.
Electrophoresis, 22(6), 1112-1118 (2001-05-19)
A fast screening method for the determination of the dissociation constants (pKa) of acidic, basic, and multivalent compounds was developed by using pressure-assisted capillary electrophoresis (PACE). External air pressure was applied to shorten the analysis time. The separation efficiency decreases
Scott E Denmark et al.
The Journal of organic chemistry, 71(4), 1668-1676 (2006-02-14)
The mechanism of the formation of substituted quinolines from anilines and alpha,beta-unsaturated ketones has been studied by the use of 13C-labeled ketones in cross-over experiments. In the reaction of doubly labeled 13C(2,4) mesityl oxide, a 100% scrambling of the label
I Z Atamna et al.
Journal of chromatography, 588(1-2), 315-320 (1991-12-27)
Expressions are formulated for the prediction of solute migration time and resolution as a function applied voltage and buffer concentration in capillary zone electrophoresis. The resolution equation assumes that solute diffusion is the only operative zone-broadening mechanism. A resolution surface
A K Malik
Journal of environmental monitoring : JEM, 2(2), 151-153 (2001-03-17)
A procedure has been developed for the determination of iron(III) dimethyldithiocarbamate by converting it into iron(II)-bathophenanthroline complex, which is then extracted into mesityl oxide in the presence of potassium perchlorate. The absorbance is then measured at 534 nm against a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service