Skip to Content
Merck
All Photos(1)

Documents

W336408

Sigma-Aldrich

Methyl trans-3-hexenoate

≥97%, FG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH=CHCH2CO2CH3
CAS Number:
Molecular Weight:
128.17
FEMA Number:
3364
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.267
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA

Assay

≥97%

refractive index

n20/D 1.4260 (lit.)

bp

169 °C (lit.)

density

0.913 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

green; fruity

SMILES string

CC\C=C\CC(=O)OC

InChI

1S/C7H12O2/c1-3-4-5-6-7(8)9-2/h4-5H,3,6H2,1-2H3/b5-4+

InChI key

XEAIHUDTEINXFG-SNAWJCMRSA-N

Related Categories

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

114.8 °F - closed cup

Flash Point(C)

46 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mengya Wang et al.
Applied microbiology and biotechnology, 102(19), 8511-8524 (2018-08-09)
Persisters represent a small bacterial population that is dormant and that survives under antibiotic treatment without experiencing genetic adaptation. Persisters are also considered one of the major reasons for recalcitrant chronic bacterial infections. Although several mechanisms of persister formation have

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service