Skip to Content
Merck
All Photos(1)

Key Documents

W335400

Sigma-Aldrich

Hexyl trans-2-butenoate

≥95%, FG

Synonym(s):

Hexyl crotonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=CHCO2(CH2)5CH3
CAS Number:
Molecular Weight:
170.25
FEMA Number:
3354
EC Number:
Council of Europe no.:
10688
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.266
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC

Assay

≥95%

refractive index

n20/D 1.438 (lit.)

density

0.885 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

green; fruity; pear; pineapple; sweet

SMILES string

[H]\C(C)=C(\[H])C(=O)OCCCCCC

InChI

1S/C10H18O2/c1-3-5-6-7-9-12-10(11)8-4-2/h4,8H,3,5-7,9H2,1-2H3/b8-4+

InChI key

MZNHUHNWGVUEAT-XBXARRHUSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

192.2 °F

Flash Point(C)

89 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service