Skip to Content
Merck
All Photos(1)

Documents

W327905

Sigma-Aldrich

Ethyl 2-mercaptopropionate

≥95%, FG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(SH)CO2C2H5
CAS Number:
Molecular Weight:
134.20
FEMA Number:
3279
EC Number:
Council of Europe no.:
11469
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
12.046

biological source

synthetic

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA

Assay

≥95%

refractive index

n20/D 1.449 (lit.)

bp

40-41 °C/1 mmHg (lit.)

density

1.031 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

musty; sulfurous

SMILES string

CCOC(=O)C(C)S

InChI

1S/C5H10O2S/c1-3-7-5(6)4(2)8/h4,8H,3H2,1-2H3

InChI key

LXXNWCFBZHKFPT-UHFFFAOYSA-N

General description

Ethyl 2-mercaptopropionate is a sulfur-containing flavor compound found in wine, Japanese soy sauce, Munster and Camembert cheeses.

Biochem/physiol Actions

Taste at 0.2 to 0.5 ppm

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

136.4 °F - closed cup

Flash Point(C)

58 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification of a powerful aroma compound in munster and camembert cheeses: ethyl 3-mercaptopropionate.
Sourabie? AM, et al.
Journal of Agricultural and Food Chemistry (2008)
Rapid Analysis of 27 Volatile Sulfur Compounds in Wine by Headspace Solid-Phase Microextraction Gas Chromatography Tandem Mass Spectrometry.
Slaghenaufi D, et al.
Food Analytical Methods, 18(3), 1-10 (2017)
Quantification and odor contribution of volatile thiols in Japanese soy sauce.
Meng Q, et al.
Food Science and Technology Research, 429-436 (2012)
Organoleptic Characteristics of Flavor Materials
Mosciano, G.
Perfumer & Flavorist, 28, 102-102 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service