Skip to Content
Merck
All Photos(1)

Key Documents

W323608

Sigma-Aldrich

Tetrahydro-4-methyl-2-(2-methyl-1-propenyl)-2H-pyran

mixture of cis and trans, ≥98%, stabilized, FG

Synonym(s):

(+)-Rose oxide, (4S)-2-(2-Methyl-1-propenyl)-4-methyltetrahydropyran

Sign Into View Organizational & Contract Pricing

Select a Size

W323608-SAMPLE-K
$94.30
1 KG
$348.00

$94.30


Check Cart for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
W323608-SAMPLE-K
$94.30
1 KG
$348.00

About This Item

Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
FEMA Number:
3236
Beilstein:
1680588
EC Number:
Council of Europe no.:
2269
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
13.037
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

$94.30


Check Cart for Availability

Request a Bulk OrderRequest more information

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

≥98%

contains

α-tocopherol, synthetic as stabilizer

refractive index

n20/D 1.452 (lit.)

bp

86 °C/20 mmHg (lit.)

density

0.866 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

geranium; green; rose

SMILES string

C[C@@H]1CCO[C@H](C1)\C=C(\C)C

InChI

1S/C10H18O/c1-8(2)6-10-7-9(3)4-5-11-10/h6,9-10H,4-5,7H2,1-3H3

InChI key

CZCBTSFUTPZVKJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

156.2 °F - closed cup

Flash Point(C)

69 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service