Skip to Content
Merck
All Photos(1)

Key Documents

W323500

Sigma-Aldrich

4,5,6,7-Tetrahydro-3,6-dimethylbenzofuran

≥95%

Synonym(s):

Menthofuran

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14O
CAS Number:
Molecular Weight:
150.22
FEMA Number:
3235
EC Number:
Council of Europe no.:
2265
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
13.035
NACRES:
NA.21
Organoleptic:
coffee; musty; earthy; pungent; nutty
grade:
Halal
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

grade

Halal

reg. compliance

FDA 21 CFR 117

Assay

≥95%

optical activity

[α]25/D +90°, c = 10 in methanol

refractive index

n20/D 1.485 (lit.)

bp

80-82 °C/13 mmHg (lit.)

density

0.97 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

coffee; musty; earthy; pungent; nutty

storage temp.

2-8°C

SMILES string

CC1CCc2c(C)coc2C1

InChI

1S/C10H14O/c1-7-3-4-9-8(2)6-11-10(9)5-7/h6-7H,3-5H2,1-2H3

InChI key

YGWKXXYGDYYFJU-UHFFFAOYSA-N

General description

4,5,6,7-Tetrahydro-3,6-dimethylbenzofuran, a monoterpene flavor compound, is one of the key constituents of peppermint oil and Euodia hortensis forma hortensis leaf oil.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

168.8 °F - closed cup

Flash Point(C)

76 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The volatile oils of Euodia hortensis forma hortensis.
Brophy JJ, et al.
Flavour and Fragrance Journal, 1(1), 17-20 (1985)
Kazuki Togashi et al.
Scientific reports, 9(1), 1704-1704 (2019-02-10)
Mint plants could theoretically serve as companion plants (CPs) that attract enemies of herbivores in tritrophic interactions. In order to explore the traits of mint volatiles as attractant cues for enemies of two-spotted spider mites, we performed Y-tube olfactometer assays
Flavor chemistry of peppermint oil (Mentha piperita L.).
Guntert M, et al.
ACS Symp. Ser., 794, 119?137-119?137 (2001)
Valerie M Kramlinger et al.
Chemico-biological interactions, 197(2-3), 87-92 (2012-04-11)
Nicotine is the primary addictive agent in tobacco products and is metabolized in humans by CYP2A6. Decreased CYP2A6 activity has been associated with decreased smoking. The extrahepatic enzyme, CYP2A13 (94% identical to CYP2A6) also catalyzes the metabolism of nicotine, but
J A Bakerink et al.
Pediatrics, 98(5), 944-947 (1996-11-01)
Hepatic and neurologic injury developed in two infants after ingestion of mint tea. Examination of the mint plants, from which the teas were brewed, indicated that they contained the toxic agent pennyroyal oil. Sera from each infant were analyzed for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service