Skip to Content
Merck
All Photos(1)

Documents

W319104

Sigma-Aldrich

2-Methylhexanoic acid

≥99%, FG

Synonym(s):

2-Methylcaproic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CH(CH3)COOH
CAS Number:
Molecular Weight:
130.18
FEMA Number:
3191
Beilstein:
1721227
EC Number:
Council of Europe no.:
582
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
8.035
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002

Assay

≥99%

refractive index

n20/D 1.422 (lit.)

bp

209-210 °C (lit.)

density

0.918 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

fatty; lard; oily; acidic

SMILES string

CCCCC(C)C(O)=O

InChI

1S/C7H14O2/c1-3-4-5-6(2)7(8)9/h6H,3-5H2,1-2H3,(H,8,9)

InChI key

CVKMFSAVYPAZTQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

2-methylhexanoic acid has been identified as one of the volatile flavor compounds in the peel and pulp of doum fruit.

Application


  • Update to RIFM fragrance ingredient safety assessment, 2-methylhexanoic acid, CAS registry number 4536-23-6.: This study updates the safety assessment of 2-methylhexanoic acid used in fragrance applications (Api et al., 2024).

  • Sustainable Biocatalytic Procedure for Obtaining New Branched Acid Esters.: This paper presents a novel biocatalytic method for synthesizing branched acid esters, utilizing 2-methylhexanoic acid as a precursor (Montiel et al., 2021).

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

134.6 °F - closed cup

Flash Point(C)

57 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Volatile Flavor Compounds of Peel and Pulp from Doum (Hyphaene thebaica L.) Fruit.
Aboshora W, et al.
American Journal of Food Science and Nutrition Research, 4(5), 165-169 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service