Skip to Content
Merck
All Photos(1)

Documents

W317209

Sigma-Aldrich

Hexyl isobutyrate

≥98%, FG

Synonym(s):

Hexyl-2-methylpropionate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCO2(CH2)5CH3
CAS Number:
Molecular Weight:
172.26
FEMA Number:
3172
EC Number:
Council of Europe no.:
646
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.478
NACRES:
NA.21

biological source

synthetic

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 178/2002

Assay

≥98%

refractive index

n20/D 1.413 (lit.)

solubility

H2O: insoluble

density

0.86 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

apple; green; fruity; pear; sweet

SMILES string

CCCCCCOC(=O)C(C)C

InChI

1S/C10H20O2/c1-4-5-6-7-8-12-10(11)9(2)3/h9H,4-8H2,1-3H3

InChI key

CYHBDKTZDLSRMY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

163.4 °F - closed cup

Flash Point(C)

73 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhilin Zhang et al.
Journal of insect science (Online), 14 (2014-12-21)
The morphological structure of the metathoracic scent glands (MTGs) in Adelphocoris suturalis was observed by utilizing scanning electron microscope (SEM). Also, the secretions of MTGs in male and female were analyzed by using gas chromatography-mass spectrometry (GC-MS). The result showed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service