Skip to Content
Merck
All Photos(1)

Key Documents

W317128

Sigma-Aldrich

cis-3-Hexenyl acetate

natural

Synonym(s):

(3Z)-3-Hexen-1-ol acetate, (3Z)-C-3-Hexenyl acetate, Leaf acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2CH2CH2CH=CHC2H5
CAS Number:
Molecular Weight:
142.20
FEMA Number:
3171
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.197

grade

Halal
Kosher
natural

refractive index

n20/D 1.427 (lit.)

bp

75-76 °C/23 mmHg (lit.)

density

0.897 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

banana; green; vegetable

SMILES string

[H]\C(CC)=C(/[H])CCOC(C)=O

InChI

1S/C8H14O2/c1-3-4-5-6-7-10-8(2)9/h4-5H,3,6-7H2,1-2H3/b5-4-

InChI key

NPFVOOAXDOBMCE-PLNGDYQASA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Natural occurrence: Apple, melon peach, pear, raspberry, strawberry, tomato and Virginia tobacco.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

134.6 °F

Flash Point(C)

57 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takeshi Shimoda et al.
Journal of chemical ecology, 31(9), 2019-2032 (2005-09-01)
We investigated volatile infochemicals possibly involved in location of the generalist predatory mite Neoseiulus californicus to plants infested with spider mites in a Y-tube olfactometer. The predators significantly preferred volatiles from lima bean leaves infested with Tetranychus urticae to uninfested
Toby J A Bruce et al.
Biology letters, 6(3), 314-317 (2009-12-25)
Plants subjected to insect attack usually increase volatile emission which attracts natural enemies and repels further herbivore colonization. Less is known about the capacity of herbivores to suppress volatiles and the multitrophic consequences thereof. In our study, the African forage
Nasser Said Mandour et al.
Journal of chemical ecology, 37(3), 267-272 (2011-02-19)
Females of the gregarious endoparasitoid Cotesia kariyai were attracted to a blend of volatiles released from corn plants infested with larvae of their host, the common armyworm (Mythimna separata). We investigated the effects of time after the last infestation (1-168
Jürgen Engelberth et al.
Molecular plant-microbe interactions : MPMI, 20(6), 707-716 (2007-06-09)
The induction of jasmonic acid (JA) is one of the major signaling events in plants in response to insect herbivore damage and leads to the activation of direct and indirect defensive measures. Green leafy volatiles, which constitute a major portion
David K Weaver et al.
Journal of economic entomology, 102(3), 1009-1017 (2009-07-21)
The wheat stem sawfly, Cephus cinctus Norton, causes severe losses in wheat grown in the northern Great Plains. Much of the affected area is planted in monoculture with wheat, Triticum aestivum L., grown in large fields alternating yearly between crop

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service