Skip to Content
Merck
All Photos(1)

Key Documents

W316709

Sigma-Aldrich

δ-Hexalactone

≥98%

Synonym(s):

δ-Caprolactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10O2
CAS Number:
Molecular Weight:
114.14
FEMA Number:
3167
EC Number:
Council of Europe no.:
641
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
10.01

biological source

synthetic

Assay

≥98%

refractive index

n20/D 1.452 (lit.)

bp

110-112 °C/15 mmHg (lit.)

density

1.037 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

sweet

SMILES string

CC1CCCC(=O)O1

InChI

1S/C6H10O2/c1-5-3-2-4-6(7)8-5/h5H,2-4H2,1H3

InChI key

RZTOWFMDBDPERY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

224.6 °F

Flash Point(C)

107 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

John F Teiber et al.
Biochemical pharmacology, 66(6), 887-896 (2003-09-10)
Human paraoxonase (PON1) was previously shown to hydrolyze over 30 different lactones (cyclic esters). In the present study purified human PON1 was found to catalyze the reverse reaction (lactonization) of a broad range of hydroxy acids. Hydroxy acid lactonization or
Dragomir I Draganov et al.
Journal of lipid research, 46(6), 1239-1247 (2005-03-18)
The paraoxonase (PON) gene family in humans has three members, PON1, PON2, and PON3. Their physiological role(s) and natural substrates are uncertain. We developed a baculovirus-mediated expression system, suitable for all three human PONs, and optimized procedures for their purification.
Hagai Tavori et al.
Bioorganic & medicinal chemistry, 16(15), 7504-7509 (2008-06-24)
Paraoxonase1 (PON1) is a HDL bound enzyme and many of the anti-atherogenic properties of HDL are attributed to PON1. The enzyme precise mechanism of protective action and its endogenous substrate remain elusive. PON1 hydrolyzes organophosphates, arylesters and lactones, whereas the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service