Skip to Content
Merck
All Photos(1)

Key Documents

W294306

Sigma-Aldrich

Propyl formate

≥97%

Synonym(s):

n-Propyl methanoate

Sign Into View Organizational & Contract Pricing

Select a Size

W294306-SAMPLE-K
$83.00
4 KG
$862.00

$83.00


Check Cart for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
W294306-SAMPLE-K
$83.00
4 KG
$862.00

About This Item

Linear Formula:
HCOOCH2CH2CH3
CAS Number:
Molecular Weight:
88.11
FEMA Number:
2943
Beilstein:
1739272
EC Number:
Council of Europe no.:
340
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.073
NACRES:
NA.21
grade:
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

$83.00


Check Cart for Availability

Request a Bulk OrderRequest more information

biological source

synthetic

Quality Level

grade

Halal
Kosher

Agency

meets purity specifications of JECFA

vapor density

3.03 (vs air)

vapor pressure

100 mmHg ( 29.5 °C)
50 mmHg ( 20 °C)

Assay

≥97%

autoignition temp.

851 °F

refractive index

n20/D 1.375 (lit.)

bp

80-81 °C (lit.)

mp

−93 °C (lit.)

density

0.904 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

berry; green; ethereal; fruity; rum; sweet

SMILES string

[H]C(=O)OCCC

InChI

1S/C4H8O2/c1-2-3-6-4-5/h4H,2-3H2,1H3

InChI key

KFNNIILCVOLYIR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

26.6 °F - closed cup

Flash Point(C)

-3 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service