Skip to Content
Merck
All Photos(1)

Key Documents

W288705

Sigma-Aldrich

Hydrocinnamaldehyde

≥95%, stabilized, FG, FCC

Synonym(s):

3-Phenylpropanal, 3-Phenylpropionaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CH2CHO
CAS Number:
Molecular Weight:
134.18
FEMA Number:
2887
Beilstein:
1071910
EC Number:
Council of Europe no.:
2013
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
5.080
NACRES:
NA.21

biological source

synthetic

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 172.515

Assay

≥95%

contains

α-tocopherol,synthetic as stabilizer
citric acid, synthetic as stabilizer

refractive index

n20/D 1.523 (lit.)

bp

97-98 °C/12 mmHg (lit.)

density

1.019 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

green; citrus; floral; peach

SMILES string

[H]C(=O)CCc1ccccc1

InChI

1S/C9H10O/c10-8-4-7-9-5-2-1-3-6-9/h1-3,5-6,8H,4,7H2

InChI key

YGCZTXZTJXYWCO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hydrocinnamaldehyde is one of the volatile constituents of cinnamon essential oil.

Biochem/physiol Actions

Taste at 20 ppm

Other Notes

Natural occurrence: Cinnamon, tomato, chicken, beer and oregano.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

203.0 °F - closed cup

Flash Point(C)

95 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Volatile constituents of cinnamon (Cinnamomum zeylanicum) oils.
Senanayake UM, et al.
Journal of Agricultural and Food Chemistry, 26(4), 822-824 (1978)
Extraction of essential oils from five cinnamon leaves and identification of their volatile compound compositions.
Wang R, et al.
Innovative Food Science & Emerging Technologies, 10(2), 289-292 (2009)
A comparison of essential oil constituents of bark, leaf, root and fruit of cinnamon (Cinnamomum zeylanicum Blum) grown in Sri Lanka.
Paranagama PA, et al.
Journal of the National Science Foundation of Sri Lanka, 29(3-4) (2010)
The arduous way to the egg: follow the nose.
Stefan Bartram et al.
Angewandte Chemie (International ed. in English), 42(39), 4729-4731 (2003-10-17)
Samir BouzBouz et al.
Organic letters, 5(11), 1995-1997 (2003-05-24)
[reaction: see text] The synthesis of (+)-strictifolione was achieved from 3-phenylproprionaldehyde by using enantioselective allyltitanations to control the stereogenic centers at C6, C4', and C6' and a cross-methathesis to control the configuration of the double bond at C1'-C2'.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service