Skip to Content
Merck
All Photos(1)

Key Documents

W287814

Sigma-Aldrich

Phenylacetic acid

natural, ≥99%

Synonym(s):

α-Toluic acid, α-Tolylic acid, Benzeneacetic acid, PAA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CO2H
CAS Number:
Molecular Weight:
136.15
FEMA Number:
2878
Beilstein:
1099647
EC Number:
Council of Europe no.:
672c
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
8.038

grade

natural

vapor density

~4 (vs air)

vapor pressure

1 mmHg ( 97 °C)

Assay

≥99%

color

white

bp

265 °C (lit.)

mp

76-78 °C (lit.)

density

1.081 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

cheese; chocolate; green; honey; meaty; musty; herbaceous; raspberry; rose; strawberry; vanilla; wine-like

SMILES string

OC(=O)Cc1ccccc1

InChI

1S/C8H8O2/c9-8(10)6-7-4-2-1-3-5-7/h1-5H,6H2,(H,9,10)

InChI key

WLJVXDMOQOGPHL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Natural occurrence: Guava, wines, papaya, raspberry, strawberry, potato, tomato, gruyere, Swiss and cheddar cheeses, mango, passion fruit, honey and tea.
Phenylacetic acid is a volatile acid that is reported to occur in balsamic vinegar and cocoa powder.

Biochem/physiol Actions

Odor at 1.0%
Taste at 50 ppm

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

269.6 °F - closed cup

Flash Point(C)

132 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solid?phase extraction method for determination of volatile compounds in traditional balsamic vinegar.
Duran Guerrero E, et al.
Journal of Separation Science, 31(16?17), 3030-3036 (2008)
Thin-layer high-vacuum distillation to isolate volatile flavour compounds of cocoa powder.
Krings U, et al.
European Food Research and Technology, 223(5), 675-675 (2006)
Gustav Nyström et al.
Nature communications, 6, 7259-7259 (2015-05-30)
Traditional thin-film energy-storage devices consist of stacked layers of active films on two-dimensional substrates and do not exploit the third dimension. Fully three-dimensional thin-film devices would allow energy storage in bulk materials with arbitrary form factors and with mechanical properties
L Ren et al.
Nanotechnology, 26(28), 284001-284001 (2015-06-27)
The prospect for spatial imaging with mass spectroscopy at the level of the cell requires new means of cell extraction to conserve molecular structure. To this aim, we demonstrate a new laser extraction process capable of extracting intact biological entities
D C Singleton et al.
Oncogene, 34(36), 4713-4722 (2014-12-09)
Hypoxia is a common feature of locally advanced breast cancers that is associated with increased metastasis and poorer survival. Stabilisation of hypoxia-inducible factor-1α (HIF1α) in tumours causes transcriptional changes in numerous genes that function at distinct stages of the metastatic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service