Skip to Content
Merck
All Photos(1)

Key Documents

W281409

Sigma-Aldrich

Octyl isovalerate

≥98%, FG

Synonym(s):

n-Octyl-3-methyl butyrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2CO2(CH2)7CH3
CAS Number:
Molecular Weight:
214.34
FEMA Number:
2814
EC Number:
Council of Europe no.:
446
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.451
NACRES:
NA.21
Organoleptic:
honey; floral; rose
grade:
FG
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

grade

FG

reg. compliance

EU Regulation 1334/2008 & 872/2012

Assay

≥98%

refractive index

n20/D 1.425 (lit.)

bp

249-251 °C (lit.)

density

0.862 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

honey; floral; rose

SMILES string

CCCCCCCCOC(=O)CC(C)C

InChI

1S/C13H26O2/c1-4-5-6-7-8-9-10-15-13(14)11-12(2)3/h12H,4-11H2,1-3H3

InChI key

FUBGRVHGQADOJI-UHFFFAOYSA-N

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service