W267406
1-(p-Methoxyphenyl)-2-propanone
≥97%
Synonym(s):
4-Methoxyphenylacetone, 4-Methoxybenzyl methyl ketone
About This Item
Recommended Products
biological source
synthetic
grade
Kosher
Assay
≥97%
refractive index
n20/D 1.525 (lit.)
bp
145 °C/25 mmHg (lit.)
density
1.067 g/mL at 25 °C (lit.)
application(s)
flavors and fragrances
Documentation
see Safety & Documentation for available documents
food allergen
no known allergens
Organoleptic
anise; spicy; fruity; balsamic; sweet
SMILES string
COc1ccc(CC(C)=O)cc1
InChI
1S/C10H12O2/c1-8(11)7-9-3-5-10(12-2)6-4-9/h3-6H,7H2,1-2H3
InChI key
WFWKNGZODAOLEO-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
215.6 °F - closed cup
Flash Point(C)
102 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service