Skip to Content
Merck
All Photos(1)

Key Documents

W252409

Sigma-Aldrich

Glucose pentaacetate

≥99%

Synonym(s):

α-D(+)-Glucose pentaacetate, 1,2,3,4,6-Penta-O-acetyl-α-D-glucopyranose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H22O11
CAS Number:
Molecular Weight:
390.34
FEMA Number:
2524
Beilstein:
98852
EC Number:
Council of Europe no.:
2524c
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
grade:
Kosher
biological source:
synthetic

biological source

synthetic

Quality Level

grade

Kosher

Assay

≥99%

optical activity

[α]20/D +98°, c = 1 in chloroform

mp

109-111 °C (lit.)

application(s)

flavors and fragrances

SMILES string

CC(=O)OC[C@H]1O[C@H](OC(C)=O)[C@H](OC(C)=O)[C@@H](OC(C)=O)[C@@H]1OC(C)=O

InChI

1S/C16H22O11/c1-7(17)22-6-12-13(23-8(2)18)14(24-9(3)19)15(25-10(4)20)16(27-12)26-11(5)21/h12-16H,6H2,1-5H3/t12-,13-,14+,15-,16+/m1/s1

InChI key

LPTITAGPBXDDGR-LJIZCISZSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1A

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jinqiang Zhang et al.
Nanotechnology, 30(3), 035401-035401 (2018-11-18)
WS2 nanotubes with carbon coatings in a core-shell structure (i.e. WS2@C) are synthesized through a facile method based on the Lewis acid-activated thioglycosylation chemistry. The obtained WS2@C shows a conformal coverage of conductive amorphous carbon on the surface of WS2
Tanya McDonald et al.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism, 40(3), 678-691 (2019-03-21)
Triheptanoin is anticonvulsant in several seizure models. Here, we investigated changes in glucose metabolism by triheptanoin interictally in the chronic stage of the pilocarpine mouse epilepsy model. After injection of [U-13C6]-glucose (i.p.), enrichments of 13C in intermediates of glycolysis and
Shigeru Tamogami et al.
Plant physiology and biochemistry : PPB, 109, 166-170 (2016-10-04)
To investigate the biotransformation pathway of airborne geraniol by Achyranthes bidentata (A. bidentata), deuterium labeled geraniol was applied with or without methyl jasmonate (MeJA), and the biosynthesized metabolites were analyzed. In A. bidentata leaves, geraniol was conjugated with glucose. The conjugate was
W J Malaisse
International journal of molecular medicine, 2(4), 383-388 (1998-12-19)
The two anomers of L-glucose pentaacetate were recently found to stimulate insulin release. The insulinotropic action of these esters cannot be attributed to the catabolism in islet cells of their glucidic or acetic moieties. The present review deals with an
Directed assembly of sub-nanometer thin organic materials with programmed-size nanopores.
Delia C Danila et al.
Angewandte Chemie (International ed. in English), 47(37), 7036-7039 (2008-08-05)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service