Skip to Content
Merck
All Photos(1)

Key Documents

W238007

Sigma-Aldrich

L-Dihydrocarvyl acetate

mixture of isomers, ≥95%

Synonym(s):

(−)-Dihydrocarvyl acetate

Sign Into View Organizational & Contract Pricing

Select a Size

W238007-SAMPLE-K
$83.00
100 G
$273.00
1 KG
$1,300.00
5 KG
$5,120.00

$83.00


Check Cart for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
W238007-SAMPLE-K
$83.00
100 G
$273.00
1 KG
$1,300.00
5 KG
$5,120.00

About This Item

Empirical Formula (Hill Notation):
C12H20O2
CAS Number:
Molecular Weight:
196.29
FEMA Number:
2380
Beilstein:
2443836
EC Number:
Council of Europe no.:
2064
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.216
NACRES:
NA.21
grade:
Fragrance grade
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

$83.00


Check Cart for Availability

Request a Bulk OrderRequest more information

biological source

synthetic

Quality Level

grade

Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
FDA 21 CFR 172.515

Assay

≥95%

optical activity

[α]/D ≤55°, neat

refractive index

n20/D 1.459 (lit.)

bp

232-234 °C (lit.)

density

0.947 g/mL at 20 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

medicinal; minty; floral; sweet

SMILES string

CC1CCC(CC1OC(C)=O)C(C)=C

InChI

1S/C12H20O2/c1-8(2)11-6-5-9(3)12(7-11)14-10(4)13/h9,11-12H,1,5-7H2,2-4H3

InChI key

TUSIZTVSUSBSQI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

194.0 °F - closed cup

Flash Point(C)

90 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service