Skip to Content
Merck
All Photos(1)

Documents

W215600

Sigma-Aldrich

Bois de rose

Synonym(s):

Rosewood oil

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2156
MDL number:
UNSPSC Code:
12164502

biological source

Aniba rosaeodora Ducke
synthetic

grade

Halal
Kosher

reg. compliance

FDA 21 CFR 117
FDA 21 CFR 182.20

refractive index

n20/D 1.464 (lit.)

bp

200-206 °C (lit.)

density

0.887 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

floral; woody; sweet

Preparation Note

Extraction method: steam distillation

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

190.0 °F - closed cup

Flash Point(C)

87.78 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service