Skip to Content
Merck
All Photos(1)

Key Documents

W214000

Sigma-Aldrich

Benzyl butyrate

≥98%, FCC, FG

Synonym(s):

Benzyl butanoate, Benzyl n-butyrate, Phenylmethyl butyrate

Sign Into View Organizational & Contract Pricing

Select a Size

W214000-SAMPLE-K
$83.00
1 KG
$109.00
5 KG
$480.00
10 KG
$747.00

$83.00


Check Cart for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
W214000-SAMPLE-K
$83.00
1 KG
$109.00
5 KG
$480.00
10 KG
$747.00

About This Item

Linear Formula:
CH3CH2CH2CO2CH2C6H5
CAS Number:
Molecular Weight:
178.23
FEMA Number:
2140
EC Number:
Council of Europe no.:
277
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.051
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

$83.00


Check Cart for Availability

Request a Bulk OrderRequest more information

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥98%

refractive index

n20/D 1.494 (lit.)

bp

240 °C (lit.)

density

1.009 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

fresh; fruity; floral

SMILES string

CCCC(=O)OCc1ccccc1

InChI

1S/C11H14O2/c1-2-6-11(12)13-9-10-7-4-3-5-8-10/h3-5,7-8H,2,6,9H2,1H3

InChI key

VONGZNXBKCOUHB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

224.6 °F - closed cup

Flash Point(C)

107 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service