Skip to Content
Merck
All Photos(1)

Documents

W213004

Sigma-Aldrich

Benzaldehyde propylene glycol acetal, mixture of isomers

≥95%

Synonym(s):

4-Methyl-2-phenyl-1,3-dioxolane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12O2
CAS Number:
Molecular Weight:
164.20
FEMA Number:
2130
EC Number:
Council of Europe no.:
2226
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
6.032

biological source

synthetic

grade

Kosher

reg. compliance

FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥95%

refractive index

n20/D 1.509 (lit.)

bp

83-85 °C/4 mmHg (lit.)

density

1.065 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Organoleptic

bitter

SMILES string

CC1COC(O1)c2ccccc2

InChI

1S/C10H12O2/c1-8-7-11-10(12-8)9-5-3-2-4-6-9/h2-6,8,10H,7H2,1H3

InChI key

CDIKGISJRLTLRA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F

Flash Point(C)

113 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service