Skip to Content
Merck
All Photos(1)

Key Documents

W211907

Sigma-Aldrich

Basil oil, comoric type

natural, FG

Synonym(s):

Ocimum basilicum

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2119
Council of Europe no.:
308n
MDL number:
UNSPSC Code:
12164502
NACRES:
NA.21
Organoleptic:
green; spicy
grade:
FG
Kosher
natural
biological source:
Ocimum basilicum L.
food allergen:
no known allergens

biological source

Ocimum basilicum L.

grade

FG
Kosher
natural

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 182.20

optical activity

[α]20/D −0.5°, neat

origin

Comores origin

refractive index

n20/D 1.515 (lit.)

bp

215 °C (lit.)

density

0.956 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

green; spicy

Biochem/physiol Actions

Taste at 10 ppm

Preparation Note

Extraction method: steam distillation

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

179.6 °F

Flash Point(C)

82 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mosciano, G.
Perfumer & Flavorist, 27, 93-93 (2002)
K J Lachowicz et al.
Letters in applied microbiology, 26(3), 209-214 (1998-05-07)
Essential oils extracted by hydrodistillation from five different varieties of Ocimum basilicum L. plants (Anise, Bush, Cinnamon, Dark Opal and a commercial sample of dried basil) were examined for antimicrobial activity against a wide range of foodborne Gram-positive and -negative
J Wan et al.
Journal of applied microbiology, 84(2), 152-158 (1998-06-20)
Basil essential oils, including basil sweet linalool (BSL) and basil methyl chavicol (BMC), were screened for antimicrobial activity against a range of Gram-positive and Gram-negative bacteria, yeasts and moulds using an agar well diffusion method. Both essential oils showed antimicrobial
Rahul Jain et al.
Drug development and industrial pharmacy, 34(4), 384-389 (2008-04-11)
The present work investigates effectiveness of basil oil, a volatile oil containing alcoholic terpenes, as a potential penetration enhancer for improved skin permeation of labetolol hydrochloride (LHCl) with reference to camphor, geraniol, thymol, and clove oil. Saturation solubilities of LHCl
Vijayalakshmi Ghosh et al.
Ultrasonics sonochemistry, 20(1), 338-344 (2012-09-08)
Basil oil (Ocimum basilicum) nanoemulsion was formulated using non-ionic surfactant Tween80 and water by ultrasonic emulsification method. Process of nanoemulsion development was optimized for parameters such as surfactant concentration and emulsification time to achieve minimum droplet diameter with high physical

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service