Skip to Content
Merck
All Photos(1)

Documents

W209000

Sigma-Aldrich

Angelica seed oil

natural, FG

Synonym(s):

Angelica oil

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2090
UNSPSC Code:
12164502
NACRES:
NA.21

biological source

Angelica archangelica

grade

FG
Kosher
natural

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

form

liquid

composition

Limonene, 2%

refractive index

n20/D 1.486

density

0.850 g/mL at 25 °C

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

woody; pepper; sweet

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 3 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

109.4 °F - closed cup

Flash Point(C)

43 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service