Skip to Content
Merck
All Photos(1)

Key Documents

U601

Sigma-Aldrich

1,11-Undecanedicarboxylic acid

94%

Synonym(s):

Brassylic acid, Tridecanedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2C(CH2)11CO2H
CAS Number:
Molecular Weight:
244.33
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
94%

Assay

94%

mp

112-114 °C (lit.)

SMILES string

OC(=O)CCCCCCCCCCCC(O)=O

InChI

1S/C13H24O4/c14-12(15)10-8-6-4-2-1-3-5-7-9-11-13(16)17/h1-11H2,(H,14,15)(H,16,17)

InChI key

DXNCZXXFRKPEPY-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Rocchiccioli et al.
Biomedical & environmental mass spectrometry, 13(6), 315-318 (1986-06-01)
The urines of children with neonatal adrenoleukodystrophy and Zellweger syndrome contained an excess of unusual even- and odd-numbered dicarboxylic acids with a chain length of from 5 to 15 carbon atoms, as well as 2-hydroxy-compounds, including 2-hydroxy-isocaproate, 2-hydroxy-glutarate and 2-hydroxy-sebacate.
Z C Wu et al.
Journal of environmental sciences (China), 13(2), 157-163 (2001-10-10)
The wastewater treatment from brassylic acid manufacturing plant using membrane bioreactor (MBR) was studied. The membrane bioreactor consisted of batch-operation biological aeration tank and ultrafiltration evaluation tank. The content of test included the affection of variation operation conditions on ultrafiltration
Wen-xun Guo et al.
Biopolymers, 74(3), 248-255 (2004-05-20)
Poly(dimer acid-brassylic acid) [P(DA-BA)] copolymers and poly(dimer acid-pentadecandioic acid) [P(DA-PA)] copolymers were prepared by melt polycondensation of the corresponding mixed anhydride prepolymers. The copolymers were characterized by Fourier transform infrared (FTIR), gel permeation chromatography (GPC), differential scanning calorimetry (DSC), wide
Xu Huang et al.
The journal of physical chemistry. B, 110(39), 19479-19486 (2006-09-29)
The gemini surfactant sodium 4,8-dioctyl-3,9-dioxo-6-hydroxy-4,8-diaza-1,11-undecanedicarboxylate (SDUC), bearing two carboxylic headgroups and two hydrocarbon chains, has been synthesized, and its self-organization characteristics at strong basic and neutral environments have been studied. The results reveal that a transition from micelles to vesicles
Garima Singh et al.
Protein science : a publication of the Protein Society, 14(2), 395-400 (2005-01-22)
This is the first evidence of a naturally bound fatty acid to a group I Phospholipase A(2) (PLA(2)) and also to a PLA(2) with Asp 49. The fatty acid identified as n-tridecanoic acid is observed at the substrate recognition site

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service