Key Documents
THS01417
ThalesNano CatCart® catalyst cartridge system, 30 mm L
nickel sponge
Synonym(s):
Heterogeneous catalysis system, safe Heterogeneous catalysis
About This Item
UNSPSC Code:
60104500
NACRES:
NB.51
Product THS01417 is not currently sold in your country. Contact Technical Service
packaging
pack of 1 ea
manufacturer/tradename
ThalesNano THS 01417
diam.
4 mm
General description
CatCarts are suitable for use only with Thalesnano H-Cube reactors.
Legal Information
CatCart is a registered trademark of ThalesNano Nanotechnology Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)