Key Documents
TEM-LC300NIUL
Sigma-Aldrich
Ultrathin Lacey Carbon Supported Nickel Grid
size 300 mesh, box of 25
Synonym(s):
LACEY CARBON FILM - NI 300 MESH ULTRA THIN 25/BX
Select a Size
1 PKG
$244.00
$244.00
Check Cart for Availability
Request a Bulk Order
Select a Size
Change View
1 PKG
$244.00
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
$244.00
Check Cart for Availability
Request a Bulk Order
packaging
box of 25
size
300 mesh
General description
Ultrathin Lacey Carbon (4-6 nm) Supported Grid: Nickel Grid, 300 mesh. This carbon coated film on a broken pattern consists of woven-mesh-like openings of different sizes and shapes. This type of pattern provides support but does not interfere when observing specimen sections.
Choose from one of the most recent versions:
Certificates of Analysis (COA)