Key Documents
TEM-FF400CU50
Sigma-Aldrich
Formvar Supported Copper Square Mesh
size 400 mesh, box of 50
Synonym(s):
FORMVAR FILM 400 MESH CU 50/BX
Select a Size
1 PKG
$133.00
$133.00
Check Cart for Availability
Request a Bulk Order
Select a Size
Change View
1 PKG
$133.00
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
$133.00
Check Cart for Availability
Request a Bulk Order
packaging
box of 50
size
400 mesh
General description
A 10 nm thin formvar film supported standard copper square mesh grids. Copper grid, 400 mesh. All of our film is laid on the shiny side of the grid.
Choose from one of the most recent versions:
Certificates of Analysis (COA)