Key Documents
TEM-FF200CUUL
Sigma-Aldrich
Ultrathin Formvar Supported Copper Square Mesh
size 200 mesh, box of 50
Synonym(s):
FORMVAR SQUARE MESH - CU 200 MESH UL 50/BX
Select a Size
1 PKG
$176.00
$176.00
Check Cart for Availability
Request a Bulk Order
Select a Size
Change View
1 PKG
$176.00
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
$176.00
Check Cart for Availability
Request a Bulk Order
packaging
box of 50
size
200 mesh
General description
A 5-6 nm ultrathin formvar film supported standard copper square mesh grids. Copper grid, 200 mesh. All of our film is laid on the shiny side of the grid.
Choose from one of the most recent versions:
Certificates of Analysis (COA)