Key Documents
TEM-FCF400AUUA
Sigma-Aldrich
Ultrathin Formvar/Carbon Supported Gold Square Mesh
size 400 mesh, box of 50
Synonym(s):
FORMVAR/CARBON MESH - AU 400 MESH UA 50/BX
Select a Size
1 PKG
$228.00
$228.00
Check Cart for Availability
Request a Bulk Order
Select a Size
Change View
1 PKG
$228.00
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
$228.00
Check Cart for Availability
Request a Bulk Order
packaging
box of 50
size
400 mesh
General description
Ultrathin 5-6nm Formvar and 1nm Carbon Supported Square Mesh Grids: Gold Grid, 400 mesh. A formvar coated grid, stabilized with evaporated carbon film. This type of coating is excellent for specimen support, especially for ultra thin sections.
Choose from one of the most recent versions:
Certificates of Analysis (COA)