Key Documents
TEM-FCF200NIUA
Sigma-Aldrich
Ultrathin Formvar/Carbon Supported Nickel Square Mesh
size 200 mesh, box of 50
Synonym(s):
FORMVAR/CARBON SQUARE MESH - NI 200 MESH UA 50/BX
Select a Size
1 PKG
$199.00
$199.00
Check Cart for Availability
Request a Bulk Order
Select a Size
Change View
1 PKG
$199.00
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
$199.00
Check Cart for Availability
Request a Bulk Order
packaging
box of 50
size
200 mesh
General description
Ultrathin 5-6nm Formvar and 1nm Carbon Supported Square Mesh Grids: Nickel Grid, 200 mesh. A formvar coated grid, stabilized with evaporated carbon film. This type of coating is excellent for specimen support, especially for ultra thin sections.
Choose from one of the most recent versions:
Certificates of Analysis (COA)