Skip to Content
Merck
All Photos(1)

Key Documents

T81205

Sigma-Aldrich

Triphenylacetic acid

99%

Synonym(s):

Tritylformic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$113.00
10 G
$207.00

$113.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$113.00
10 G
$207.00

About This Item

Linear Formula:
(C6H5)3CCO2H
CAS Number:
Molecular Weight:
288.34
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$113.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

mp

270-273 °C (lit.)

SMILES string

OC(=O)C(c1ccccc1)(c2ccccc2)c3ccccc3

InChI

1S/C20H16O2/c21-19(22)20(16-10-4-1-5-11-16,17-12-6-2-7-13-17)18-14-8-3-9-15-18/h1-15H,(H,21,22)

InChI key

DCYGAPKNVCQNOE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Hazard Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

S Fournel et al.
Biochimica et biophysica acta, 883(2), 190-196 (1986-09-04)
Bilirubin UDPglucuronosyltransferase of rat or human liver microsomes was inhibited, in vitro, by triphenylacetic acid and by structurally related arylcarboxylic acids. This inhibition appeared to be competitive towards bilirubin, and mixed-type towards UDPglucuronic acid. A decrease in the number of
A D Blackwood et al.
Biochimica et biophysica acta, 1206(2), 161-165 (1994-06-12)
Combinations of triisooctylamine with its hydrochloride, or of triphenylacetic acid with its Na+ salt, can function as buffers for use during biocatalysis in organic media. They can control the pH of an adjacent aqueous phase, even though both forms of
Grant A McNaughton-Smith et al.
Journal of medicinal chemistry, 51(4), 976-982 (2008-02-01)
Sickle cell disease (SCD) is a hereditary condition characterized by deformation of red blood cells (RBCs). This phenomenon is due to the presence of abnormal hemoglobin that polymerizes upon deoxygenation. This effect is exacerbated when dehydrated RBCs experience a loss

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service